Learn More
Bio X Cell InVivoMAb TIM-4, F31-5G3, Rat IgG1, 5 MG
Supplier: Bio X Cell BE0344R005MG
The F31-5G3 monoclonal antibody reacts specifically with mouse T cell immunoglobulin and mucin domain 4 (TIM-4), a phosphatidylserine-binding receptor. TIM-4 is expressed on antigen-presenting cells, enhancing apoptotic cell engulfment and regulating T cell proliferation.
- Formulation: PBS, pH 7.0, free of stabilizers or preservatives
- Endotoxin level: ≤1 EU/mg (≤0.001 EU/μg), determined by LAL assay
- Purity: ≥95%, verified by SDS-PAGE
- Sterility: 0.2 μm filtration
- Production: Purified from cell culture supernatant in an animal-free facility
- Purification method: Protein G
- Molecular weight: Approximately 150 kDa
- Storage: Store at 4°C at stock concentration; avoid freezing
- Conjugation: Unconjugated (conjugation services available)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.