Learn More
Bio X Cell InVivoMAb anti-mouse CD40, Clone FGK4.5/FGK45, Rat IgG2a, 1 MG
Supplier: Bio X Cell BE00162R001MG
The FGK4.5 monoclonal antibody targets mouse CD40 (Bp50), a 48 kDa transmembrane glycoprotein of the TNFR superfamily. CD40 is expressed on antigen-presenting cells (APCs) and other cell types, acting as a costimulatory molecule vital for APC activation, B cell activation, differentiation, and Ig isotype switching. This agonistic antibody activates CD40 expressing APCs, promotes anti-tumor T cell responses, and can inhibit CD40/CD154 interaction.
- Isotype: Rat IgG2a
- Immunogen: Recombinant mouse CD40 fusion protein
- Applications: In vivo CD40 activation, in vitro B cell stimulation
- Formulation: PBS, pH 7.0, no preservatives
- Endotoxin: 1EU/mg
- Purity: 95%
- Sterility: 0.2 m filtration
- Storage: 4C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.