Learn More
Bio X Cell Fusion Protein, Flt-3L-Ig, Mouse, Human, 1 MG
Supplier: Bio X Cell BE0098R001MG
Flt-3L (FMS-related Tyrosine Kinase 3 Ligand) is an endogenous cytokine and growth factor crucial for the development of conventional and plasmacytoid dendritic cells (cDCs and pDCs). This recombinant Flt-3L-Ig fusion protein, composed of human Flt-3L fused to the Fc portion of human IgG1, activates Flt3 signaling and induces DC population expansion, with reported in vivo stimulation in mice.
- Clone: Flt-3L-Ig (hum/hum)
- Reactivities: Mouse, human
- Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95% determined by SDS-PAGE
- Sterility: 0.2 μm filtered
- Purification: Protein A
- Storage: Store at stock concentration at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.