Learn More
Bio X Cell InVivoMAb Flt-3L-Ig, Human Mouse Reactivity, 50 MG
Supplier: Bio X Cell BE0098R050MG
Flt-3L (FMS-related Tyrosine Kinase 3 Ligand) is an endogenous protein that functions as a cytokine and growth factor. Flt-3L is crucial for the development of conventional dendritic cells (cDCs) and plasmacytoid dendritic cells (pDCs). Recombinant Flt-3L-Ig is a fusion protein consisting of human Flt-3L fused to the Fc portion of human IgG1. This fusion protein is useful for activating Flt3 signaling and inducing the expansion of DC populations. Human Flt-3L-Ig is frequently reported to stimulate Flt3 signaling in vivo in mice.
- Endotoxin ≤1EU/mg (≤0.001EU/μg), determined by LAL assay
- Purity ≥95%, determined by SDS-PAGE
- Sterility 0.2 μm filtration
- Purified from cell culture supernatant in an animal-free facility
- Purification by Protein A
- Formulated in PBS, pH 7.0, contains no stabilizers or preservatives
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.