Learn More
Bio X Cell Antibody, anti-mouse CD70, Clone FR70, Rat IgG2b Isotype, 1 MG
Supplier: Bio X Cell BE0022R001MG
The FR70 monoclonal antibody targets mouse CD70, a 30-33 kDa type II transmembrane glycoprotein and a member of the TNF superfamily. This antibody blocks the interaction between CD70, which is expressed on activated mouse T and B lymphocytes and dendritic cells, and its ligand CD27. This interaction is known to promote T and B cell cross-stimulation, B cell proliferation, and immunoglobulin production.
- Monoclonal antibody, clone FR70
- Reacts with mouse CD70
- Blocks CD70 binding to CD27
- Isotype is rat IgG2b, kappa
- Purity of 95% or greater
- Endotoxin level is 1 EU/mg or less
- Formulated in PBS, pH 7.0, without stabilizers or preservatives
- Sterile via 0.2 m filtration
- Purified in an animal-free facility using protein G
- Reported applications include in vivo CD70 blockade, in vitro CD70 blockade, and flow cytometry
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.