Learn More
Bio X Cell Monoclonal Antibody, Anti-Mouse IL-23 (p19), 25 MG
Supplier: Bio X Cell BE0313R025MG
The G23-8 monoclonal antibody specifically reacts with the p19 subunit of mouse IL-23, a heterodimeric cytokine involved in immune response. This antibody neutralizes IL-23 bioactivity without affecting IL-12 p70 bioactivity, making it suitable for studies on autoimmune diseases and infections.
- Clone: G23-8
- Reactivities: mouse
- Isotype: rat IgG1, κ
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Purification: protein G
- Molecular weight: 150 kDa
- Storage: 4°C, do not freeze
- Applications: in vivo IL-23p19 neutralization, Western blot
- Immunogen: insect cell-expressed, recombinant mouse IL-23 heterodimer
- Conjugation: unconjugated
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.