Learn More
Bio X Cell anti-human CD40, Clone G28.5, IgG1, 1 MG
Supplier: Bio X Cell BE0189R001MG
The InVivoMAb anti-human CD40, Clone G28.5, is an agonistic monoclonal antibody that reacts with human CD40, a 48 kDa type I transmembrane glycoprotein. This antibody activates CD40 expressing antigen-presenting cells (APCs) and promotes anti-tumor T cell responses.
- Reacts with human CD40
- Agonistic antibody that activates CD40 expressing APCs
- Purified from cell culture supernatant in an animal-free facility
- Purity ≥95% by SDS-PAGE
- Endotoxin ≤1 EU/mg (≤0.001 EU/μg) by LAL assay
- Sterility by 0.2 μm filtration
- Clone G28.5, mouse IgG1, κ isotype
- Target: human CD40
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.