Learn More
Bio X Cell anti-mouse CD326, G8.8, Rat IgG2a, 1 MG
Supplier: Bio X Cell BE0346R001MG
The G8.8 monoclonal antibody reacts with CD326, also known as EpCAM, a 40-42 kDa cell-surface type 1 transmembrane glycoprotein. It is expressed on most epithelial cells and plays a role in cell-cell adhesion and as a potential growth factor receptor.
- Host: Rat
- Target species: Mouse CD326 (EpCAM)
- Isotype: Rat IgG2a, κ
- Molecular weight: 150 kDa
- Purity: ≥95% (determined by SDS-PAGE)
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Purification: Protein G from animal-free facility
- Sterility: 0.2 μm filtration
- Storage: Store at 4°C, do not freeze
- Applications: Immunohistochemistry (frozen), Immunofluorescence, Flow cytometry, Western blot
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.