Learn More
Bio X Cell Antibody, anti-mouse CD326, Rat IgG2a, 5 MG
Supplier: Bio X Cell BE0346R005MG
The G8.8 monoclonal antibody reacts with CD326, also known as EpCAM, a cell-surface glycoprotein involved in cell-cell adhesion and a potential growth factor receptor. This antibody is used as a target for immunotherapy treatment of human carcinomas.
- Isotype: Rat IgG2a, κ
- Purity: ≥95%
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Sterility: 0.2 μm filtration
- Production: Purified from cell culture supernatant in an animal-free facility
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at stock concentration at 4°C, do not freeze
- Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
- Reactivities: Mouse
- Unconjugated
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.