Learn More
Bio X Cell InVivoMAb, CD326, G8.8, Rat IgG2a, 50 MG
Supplier: Bio X Cell BE0346R050MG
The G8.8 monoclonal antibody specifically targets CD326 (EpCAM), a 40-42 kDa cell-surface glycoprotein expressed on most epithelial cells and certain immune cells, mediating cell-cell adhesion and acting as a potential growth factor receptor.
- Isotype: Rat IgG2a, κ
- Purification method: Protein G
- Formulation: PBS, pH 7.0; contains no stabilizers or preservatives
- Endotoxin level: ≤1 EU/mg
- Purity: ≥95% by SDS-PAGE
- Sterility: 0.2 μm filtration
- Molecular weight: 150 kDa
- Storage conditions: Store at 4°C; do not freeze
- Reported applications: Immunohistochemistry (frozen), immunofluorescence, flow cytometry, western blot
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.