Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell Antibody, CD86, GL-1, Rat IgG2a, 1 MG

Catalog No. 501528254
Encompass_Preferred

The GL-1 monoclonal antibody reacts with mouse CD86, also known as B7-2. CD86 is an 80 kDa Ig superfamily member expressed by activated T and B cells, macrophages, and dendritic cells. This antibody blocks CD86 in vivo and inhibits the priming of cytotoxic T cells.

  • Clone: GL-1
  • Reactivity: Mouse
  • Isotype: Rat IgG2a, κ
  • Purification method: Protein G
  • Formulation: PBS, pH 7.0, no stabilizers or preservatives
  • Endotoxin: ≤1EU/mg (≤0.001EU/μg)
  • Purity: ≥95%
  • Sterility: 0.2 μm filtration
  • Molecular weight: 150 kDa
  • Storage: Store at stock concentration at 4°C, do not freeze
  • Applications: In vivo CD86 blockade, flow cytometry
  • Immunogen: LPS-activated CBA/Ca mouse splenic B cells
  • Conjugation: Unconjugated

Catalog No. 50-152-8254 Supplier Bio X Cell Supplier No. BE0025R001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The GL-1 monoclonal antibody reacts with mouse CD86, also known as B7-2. CD86 is an 80 kDa Ig superfamily member expressed by activated T and B cells, macrophages, and dendritic cells. This antibody blocks CD86 in vivo and inhibits the priming of cytotoxic T cells.

  • Clone: GL-1
  • Reactivity: Mouse
  • Isotype: Rat IgG2a, κ
  • Purification method: Protein G
  • Formulation: PBS, pH 7.0, no stabilizers or preservatives
  • Endotoxin: ≤1EU/mg (≤0.001EU/μg)
  • Purity: ≥95%
  • Sterility: 0.2 μm filtration
  • Molecular weight: 150 kDa
  • Storage: Store at stock concentration at 4°C, do not freeze
  • Applications: In vivo CD86 blockade, flow cytometry
  • Immunogen: LPS-activated CBA/Ca mouse splenic B cells
  • Conjugation: Unconjugated

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.