Learn More
Bio X Cell Isotype Control, Human IgG2, Unconjugated, 1 MG
Supplier: Bio X Cell BE0301R001MG
This human IgG2 isotype control antibody is purified from human myeloma serum for use as a non-targeting control in various in vitro and in vivo studies. It also serves as a negative control in diagnostic applications such as ELISA, Western blot, immunofluorescence, immunohistochemistry, immunoprecipitation, and flow cytometry. For research use only.
- Purification method: Protein A
- Purity: ≥95% (determined by SDS-PAGE)
- Endotoxin: <2EU/mg (<0.002EU/μg)
- Sterility: 0.2 μm filtration
- Molecular weight: 150 kDa
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Storage: Store at stock concentration at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.