Learn More
Bio X Cell InVivoMAb Anti-Human CD32 (FcgammaRIIA), 1 MG
Catalog No. 501528415
Encompass_Preferred
Bio X Cell InVivoMAb anti-human CD32 (FcγRIIA) is a mouse monoclonal antibody (IgG2b) produced using a human K562 leukemia cell line immunogen. It is supplied in a 1 mg size.
  • Designed for in vivo FcγRIIA blockade in humanized mice
  • Suitable for in vitro FcγRIIA blockade studies
  • Validated for ELISA applications
  • Compatible with flow cytometry
  • Targets human CD32 (FcγRIIA)
  • Produced in mouse host
  • Isotype IgG2b
  • Immunized with human K562 leukemia cell line
Catalog No. 50-152-8415 Supplier Bio X Cell Supplier No. BE0224R001MG
Bio X Cell InVivoMAb anti-human CD32 (FcγRIIA) is a mouse monoclonal antibody (IgG2b) produced using a human K562 leukemia cell line immunogen. It is supplied in a 1 mg size.
  • Designed for in vivo FcγRIIA blockade in humanized mice
  • Suitable for in vitro FcγRIIA blockade studies
  • Validated for ELISA applications
  • Compatible with flow cytometry
  • Targets human CD32 (FcγRIIA)
  • Produced in mouse host
  • Isotype IgG2b
  • Immunized with human K562 leukemia cell line