Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell Antibody, J116, PD-1, IgG1, 1 MG

Catalog No. 501528387
Encompass_Preferred

The J116 monoclonal antibody reacts with human PD-1 (CD279), a 50-55 kDa cell surface receptor that plays key roles in peripheral tolerance and the prevention of autoimmune disease. The antibody is reported to inhibit PD-1 signal transduction.

  • Clone: J116
  • Target: Human PD-1 (CD279)
  • Isotype: Mouse IgG1, κ
  • Reported applications: In vitro PD-1 neutralization, in vivo PD-1 blockade in humanized mice, in vitro Organoids/Organ-on-Chip
  • Molecular weight: 150 kDa
  • Purity: ≥95% (Determined by SDS-PAGE)
  • Endotoxin: ≤1EU/mg (≤0.001EU/μg)
  • Sterility: 0.2 μm filtered
  • Purification: Protein G
  • Formulation: PBS, pH 7.0; no stabilizers or preservatives
  • Storage: Store at 4°C; do not freeze

Catalog No. 50-152-8387 Supplier Bio X Cell Supplier No. BE0188R001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The J116 monoclonal antibody reacts with human PD-1 (CD279), a 50-55 kDa cell surface receptor that plays key roles in peripheral tolerance and the prevention of autoimmune disease. The antibody is reported to inhibit PD-1 signal transduction.

  • Clone: J116
  • Target: Human PD-1 (CD279)
  • Isotype: Mouse IgG1, κ
  • Reported applications: In vitro PD-1 neutralization, in vivo PD-1 blockade in humanized mice, in vitro Organoids/Organ-on-Chip
  • Molecular weight: 150 kDa
  • Purity: ≥95% (Determined by SDS-PAGE)
  • Endotoxin: ≤1EU/mg (≤0.001EU/μg)
  • Sterility: 0.2 μm filtered
  • Purification: Protein G
  • Formulation: PBS, pH 7.0; no stabilizers or preservatives
  • Storage: Store at 4°C; do not freeze

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.