Learn More
Bio X Cell Antibody, anti-mouse IL-1 R JAMA-147, 1 MG
Supplier: Bio X Cell BE0256R001MG
The JAMA-147 monoclonal antibody targets mouse IL-1 receptor (IL-1 R) type 1, also known as CD121a. This 80 kDa transmembrane glycoprotein is expressed on T cells, thymocytes, dendritic cells, fibroblasts, vascular endothelial cells, epithelial cells, and neural cells, mediating IL-1 biological responses upon ligand binding.
- Isotype: Armenian Hamster IgG, κ
- Immunogen: extracellular domain of mouse IL-1 R type 1
- Formulation: PBS, pH 6.0, 0.01% Tween; contains no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95%, determined by SDS-PAGE
- Sterility: 0.2 μm filtration
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: store at stock concentration at 4°C; do not freeze
- Applications: in vivo and in vitro IL-1 R blockade
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.