Learn More
Bio X Cell Antibody, anti-mouse CD3, Clone KT3, Unconjugated, 1 MG
Supplier: Bio X Cell BE0261R001MG
This monoclonal antibody, clone KT3, targets mouse CD3, a 20 kDa transmembrane protein integral to the T cell receptor (TCR) complex. Expressed on T lymphocytes, NK-T cells, and developing thymocytes, CD3 is crucial for TCR signaling, T lymphocyte activation, and antigen recognition. The antibody functions by binding and stimulating the TCR, thereby inducing T lymphocyte activation.
- Reactivities: mouse
- Isotype: rat IgG2a
- Clone: KT3
- Molecular weight: 150 kDa
- Conjugation: unconjugated
- Formulation: PBS, pH 7.0 (no stabilizers or preservatives)
- Endotoxin: 1 EU/mg
- Purity: 95%
- Storage: 4C (do not freeze)
- Applications: in vitro T cell negative selection, stimulation, immunofluorescence
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.