Learn More
Bio X Cell Antibody, L243, anti-Human/Monkey HLA-DR, 1 MG
Supplier: Bio X Cell BE0306R001MG
The L243 monoclonal antibody reacts with human and monkey MHC class II, HLA-DR. It is expressed primarily on antigen presenting cells and is critical for the presentation of peptides to CD4+ T cells.
- Validated for in vitro blocking of MHC class II HLA-DR, HLA class II binding assay, and MHC class II HLA-DR expressing cell negative selection
- Suitable for Western blot, flow cytometry, and immunopeptidomics applications
- Purified by Protein G, with ≥95% purity determined by SDS-PAGE
- Formulated in PBS, pH 7.0, containing no stabilizers or preservatives
- Sterility ensured by 0.2 μm filtration
- Endotoxin level ≤1EU/mg (≤0.001EU/μg)
- Store at stock concentration at 4°C; do not freeze
- Produced in an animal-free facility with a molecular weight of 150 kDa
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.