Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell Antibody, HLA-DR, Clone L243, 5 MG

Catalog No. 501528489
Encompass_Preferred

The L243 monoclonal antibody reacts with human and monkey MHC class II, HLA-DR. HLA-DR is a transmembrane glycoprotein expressed primarily on antigen-presenting cells, critical for presentation of peptides to CD4+ T cells.

  • Purified using Protein G from animal-free cell culture supernatant
  • Purity ≥95%, determined by SDS-PAGE
  • Formulated in PBS, pH 7.0, without stabilizers or preservatives
  • Endotoxin ≤1EU/mg, determined by LAL assay
  • Store at 4°C; do not freeze
  • Applications: in vitro blocking, HLA class II binding assay, negative selection, Western blot, flow cytometry, immunopeptidomics
  • Isotype: Mouse IgG2a, κ
  • Immunogen: Human lymphoblastoid B cell line RPMI 8866.9
  • Sterility: 0.2 μm filtration
  • Molecular weight: 150 kDa

Catalog No. 50-152-8489 Supplier Bio X Cell Supplier No. BE0306R005MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The L243 monoclonal antibody reacts with human and monkey MHC class II, HLA-DR. HLA-DR is a transmembrane glycoprotein expressed primarily on antigen-presenting cells, critical for presentation of peptides to CD4+ T cells.

  • Purified using Protein G from animal-free cell culture supernatant
  • Purity ≥95%, determined by SDS-PAGE
  • Formulated in PBS, pH 7.0, without stabilizers or preservatives
  • Endotoxin ≤1EU/mg, determined by LAL assay
  • Store at 4°C; do not freeze
  • Applications: in vitro blocking, HLA class II binding assay, negative selection, Western blot, flow cytometry, immunopeptidomics
  • Isotype: Mouse IgG2a, κ
  • Immunogen: Human lymphoblastoid B cell line RPMI 8866.9
  • Sterility: 0.2 μm filtration
  • Molecular weight: 150 kDa

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.