Learn More
Bio X Cell Anti-Mouse 4-1BB, LOB12.3, Rat IgG1, 1 MG
Supplier: Bio X Cell BE0169R001MG
The LOB12.3 monoclonal antibody reacts with mouse 4-1BB, a TNF receptor superfamily member also known as CD137. This antibody provides costimulatory signals to T cells and NK cells, stimulating 4-1BB signaling and delaying tumor growth in vivo when combined with immune checkpoint inhibitors.
- Reacts with mouse 4-1BB (CD137)
- Clone: LOB12.3
- Isotype: Rat IgG1, κ
- Immunogen: Mouse CD137 human Fc fusion protein
- Reported applications: in vivo activation of 4-1BB
- Purity: ≥95% (SDS-PAGE)
- Endotoxin: ≤1EU/mg
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.