Learn More
Bio X Cell Rat IgG2b Isotype Control, LTF-2, 1 MG
Supplier: Bio X Cell BE0090R001MG
The LTF-2 monoclonal antibody is an InVivoMAb rat IgG2b isotype control. It reacts with keyhole limpet hemocyanin (KLH) and is suitable as an isotype-matched control for rat IgG2b antibodies in various in vivo and in vitro applications.
- Purified by Protein G
- Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
- Endotoxin ≤1 EU/mg
- Purity ≥95% by SDS-PAGE
- Sterility by 0.2 μm filtration
- Produced in an animal-free facility
- Molecular weight: 150 kDa
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.