Learn More
Bio X Cell Anti-Mouse CD24, M1/69, Rat IgG2b, 1 MG
Supplier: Bio X Cell BE0360R001MG
The M1/69 monoclonal antibody reacts with mouse CD24, also known as heat stable antigen (HSA), a 35-50 kDa glycosylated membrane protein. This antibody is utilized in research for resolving stages of B cell lymphopoiesis and investigating CD24's role in cell migration, invasion, and proliferation in cancer.
- Clone: M1/69
- Reactivity: Mouse
- Isotype: Rat IgG2b, κ
- Conjugation: Unconjugated
- Immunogen: C57BL/10 mouse splenic T cells
- Reported applications: In vivo administration, immunohistochemistry (frozen/paraffin), immunofluorescence, flow cytometry
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Purity: ≥95% by SDS-PAGE
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purification: Protein G
- Sterility: 0.2 μm filtration
- Storage: Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.