Learn More
Bio X Cell Anti-Mouse CD24, Clone M1/69, IgG2b, 100 MG
Supplier: Bio X Cell BE0360R100MG
The M1/69 monoclonal antibody reacts with mouse CD24, also known as heat stable antigen (HSA), a 35-50 kDa glycosylated membrane protein. This antibody is purified from cell culture supernatant in an animal-free facility.
- Isotype: Rat IgG2b, κ
- Specificity: Mouse CD24
- Conjugation: Unconjugated
- Immunogen: C57BL/10 mouse splenic T cells
- Applications: In vivo administration, immunohistochemistry (frozen), immunohistochemistry (paraffin), immunofluorescence, and flow cytometry
- Purity: ≥95% by SDS-PAGE
- Endotoxin: ≤1 EU/mg
- Storage: Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.