Learn More
Bio X Cell InVivoMAb CD24 (M1/69), Rat IgG2b, 5 MG
Supplier: Bio X Cell BE0360R005MG
The M1/69 monoclonal antibody reacts with mouse CD24 (heat stable antigen, HSA), a 35-50 kDa glycosylated membrane protein expressed on various immune cells. It is used to resolve stages of B cell lymphopoiesis and is relevant in cancer research due to its role in cell migration and proliferation.
- Supports in vivo administration, immunohistochemistry (frozen and paraffin), immunofluorescence, and flow cytometry.
- Purified by Protein G from animal-free cell culture supernatant.
- Formulated in PBS, pH 7.0, without stabilizers or preservatives.
- Store at 4°C; do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.