Learn More
Bio X Cell Antibody, anti-CD11b, Mouse/Human, M1/70, Unconjugated, 1 MG
Supplier: Bio X Cell BE0007R001MG
This Bio X Cell monoclonal rat antibody targets Integrin alpha-M (CD11b) and is suitable for various immunological applications including flow cytometry, immunohistochemistry, and in vivo experiments. It is supplied unconjugated for versatile use in research.
- Target: Integrin alpha-M (Mouse)
- Clone: Monoclonal (M1/70)
- Host species: Rat
- Reactivity: Homo sapiens (Human), Mus musculus (House mouse), Cricetulus griseus (Chinese hamster)
- Applications: Flow cytometry/Cell sorting (FC/FACS), In vivo experiments, Immunodepletion (Immu-depl), Immunohistochemistry (IHC), Immunohistochemistry-immunofluorescence (IHC-IF), Magnetic cell separation (MACS), Neutralization experiments
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.