Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell InVivoMAb anti-CD11b, M1/70, rat IgG2b, 50 MG

Catalog No. NC3343242
Encompass_Preferred

The M1/70 monoclonal antibody reacts with mouse and human CD11b, a 170 kDa membrane glycoprotein also known as integrin alpha M (ITGAM). CD11b belongs to the integrin alpha family and is primarily expressed on granulocytes and monocytes/macrophages but also expressed on dendritic cells, NK cells, and subsets of T and B cells. CD11b and CD18 combine to form Mac-1. Mac-1 functions as a complement receptor as well as a receptor for fibrinogen, factor X, and ICAM1.

  • In vivo CD11b neutralization
  • ILC2 cell purification
  • Flow cytometry
  • Purity: ≥95%, determined by SDS-PAGE
  • Endotoxin: ≤1EU/mg (≤0.001EU/μg)
  • Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
  • Purification: Protein G
  • Sterility: 0.2 μm filtered

Catalog No. NC3343242 Supplier Bio X Cell Supplier No. BE0007R050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The M1/70 monoclonal antibody reacts with mouse and human CD11b, a 170 kDa membrane glycoprotein also known as integrin alpha M (ITGAM). CD11b belongs to the integrin alpha family and is primarily expressed on granulocytes and monocytes/macrophages but also expressed on dendritic cells, NK cells, and subsets of T and B cells. CD11b and CD18 combine to form Mac-1. Mac-1 functions as a complement receptor as well as a receptor for fibrinogen, factor X, and ICAM1.

  • In vivo CD11b neutralization
  • ILC2 cell purification
  • Flow cytometry
  • Purity: ≥95%, determined by SDS-PAGE
  • Endotoxin: ≤1EU/mg (≤0.001EU/μg)
  • Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
  • Purification: Protein G
  • Sterility: 0.2 μm filtered

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.