Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell InVivoMAb Antibody, LFA-1 (CD11a), M17/4 Clone, Rat IgG2a, 1 MG

Catalog No. 501528234
Encompass_Preferred

The M17/4 monoclonal antibody targets mouse LFA-1 (lymphocyte function-associated antigen 1 alpha), also known as integrin alpha L chain and CD11a. LFA-1 combines with CD18 to form LFA-1, a 180 kDa glycoprotein belonging to the integrin family. LFA-1 is found on the surface of all leukocytes and plays a critical role in leukocyte intercellular adhesion and lymphocyte costimulatory signaling.

  • Targets mouse LFA-1 (CD11a)
  • Clone M17/4
  • Isotype: Rat IgG2a,
  • Recommended for in vivo LFA-1 neutralization
  • Recommended for flow cytometry
  • Formulated in PBS, pH 7.0, without stabilizers or preservatives
  • Endotoxin level: 1 EU/mg
  • Purity: 95% by SDS-PAGE
  • Sterility: 0.2 m filtered
  • Produced in an animal-free facility
  • Purified by Protein G
  • Molecular weight: 150 kDa

Catalog No. 50-152-8234 Supplier Bio X Cell Supplier No. BE0006R001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The M17/4 monoclonal antibody targets mouse LFA-1 (lymphocyte function-associated antigen 1 alpha), also known as integrin alpha L chain and CD11a. LFA-1 combines with CD18 to form LFA-1, a 180 kDa glycoprotein belonging to the integrin family. LFA-1 is found on the surface of all leukocytes and plays a critical role in leukocyte intercellular adhesion and lymphocyte costimulatory signaling.

  • Targets mouse LFA-1 (CD11a)
  • Clone M17/4
  • Isotype: Rat IgG2a,
  • Recommended for in vivo LFA-1 neutralization
  • Recommended for flow cytometry
  • Formulated in PBS, pH 7.0, without stabilizers or preservatives
  • Endotoxin level: 1 EU/mg
  • Purity: 95% by SDS-PAGE
  • Sterility: 0.2 m filtered
  • Produced in an animal-free facility
  • Purified by Protein G
  • Molecular weight: 150 kDa

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.