Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell Monoclonal Antibody, anti-mouse/human IL-7, M25, 1 MG

Catalog No. 501528276
Encompass_Preferred

The M25 monoclonal antibody targets mouse and human IL-7, a 20-28 kDa hematopoietic growth factor. It promotes lymphoid cell differentiation, proliferation, and homeostasis, and is used for IL-7 neutralization or administration.

  • Neutralizes IL-7 bioactivity; useful for cytokine administration and prolonged in vivo availability when complexed.
  • Isotype: Mouse IgG2b.
  • Formulation: PBS, pH 7.0; no stabilizers or preservatives.
  • Endotoxin: ≤1EU/mg (≤0.001EU/μg).
  • Purity: ≥95% by SDS-PAGE; 0.2 μm filtered.
  • Production: Purified from animal-free cell culture via Protein G.
  • Molecular weight: 150 kDa.
  • Reactivities: Mouse, human.
  • Clone: M25.
  • Immunogen: Recombinant human IL-7.
  • Storage: 4°C stock concentration; do not freeze.
  • Applications: In vivo IL-7 neutralization and receptor stimulation.

Catalog No. 50-152-8276 Supplier Bio X Cell Supplier No. BE0048R001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The M25 monoclonal antibody targets mouse and human IL-7, a 20-28 kDa hematopoietic growth factor. It promotes lymphoid cell differentiation, proliferation, and homeostasis, and is used for IL-7 neutralization or administration.

  • Neutralizes IL-7 bioactivity; useful for cytokine administration and prolonged in vivo availability when complexed.
  • Isotype: Mouse IgG2b.
  • Formulation: PBS, pH 7.0; no stabilizers or preservatives.
  • Endotoxin: ≤1EU/mg (≤0.001EU/μg).
  • Purity: ≥95% by SDS-PAGE; 0.2 μm filtered.
  • Production: Purified from animal-free cell culture via Protein G.
  • Molecular weight: 150 kDa.
  • Reactivities: Mouse, human.
  • Clone: M25.
  • Immunogen: Recombinant human IL-7.
  • Storage: 4°C stock concentration; do not freeze.
  • Applications: In vivo IL-7 neutralization and receptor stimulation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.