Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell anti-mouse CD103, Clone M290, Unconjugated, 1 MG

Catalog No. 501528255
Encompass_Preferred

The M290 monoclonal antibody reacts with mouse CD103 (integrin αE), a protein expressed on intraepithelial lymphocyte (IEL) T cells and some peripheral regulatory T cells. It is reported to neutralize CD103 in vivo.

  • Host: Rat
  • Isotype: Rat IgG2a, κ
  • Purification: Protein G
  • Formulation: PBS, pH 7.0. Contains no stabilizers or preservatives.
  • Storage: Store at stock concentration at 4°C. Do not freeze.
  • Applications: In vivo CD103 neutralization, immunofluorescence, flow cytometry
  • Purity: ≥95%
  • Endotoxin: ≤1EU/mg
  • Sterility: 0.2 μm filtration
  • Production: Purified from cell culture supernatant in an animal-free facility

Catalog No. 50-152-8255 Supplier Bio X Cell Supplier No. BE0026R001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The M290 monoclonal antibody reacts with mouse CD103 (integrin αE), a protein expressed on intraepithelial lymphocyte (IEL) T cells and some peripheral regulatory T cells. It is reported to neutralize CD103 in vivo.

  • Host: Rat
  • Isotype: Rat IgG2a, κ
  • Purification: Protein G
  • Formulation: PBS, pH 7.0. Contains no stabilizers or preservatives.
  • Storage: Store at stock concentration at 4°C. Do not freeze.
  • Applications: In vivo CD103 neutralization, immunofluorescence, flow cytometry
  • Purity: ≥95%
  • Endotoxin: ≤1EU/mg
  • Sterility: 0.2 μm filtration
  • Production: Purified from cell culture supernatant in an animal-free facility

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.