Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell Monoclonal Antibody, Anti-mouse IFNAR-1, IgG1, 1 MG

Supplier:  Bio X Cell BE0241R001MG

Encompass_Preferred

The MAR1-5A3 monoclonal antibody targets mouse IFNAR-1, inhibiting Type I IFN receptor signaling in vitro and in vivo.

  • Target: IFNAR-1 (IFN alpha/beta receptor subunit 1)
  • Clone: MAR1-5A3
  • Isotype: Mouse IgG1, κ
  • Reactivity: Mouse
  • Applications: In vivo IFNAR-1 blockade, in vitro IFNAR-1 blockade, Western blot, Flow Cytometry, ELISA
  • Formulation: PBS, pH 7.0, no stabilizers or preservatives
  • Purity: ≥95% by SDS-PAGE
  • Endotoxin: ≤1EU/mg
  • Sterility: 0.2 μm filtration
  • Purification: Protein G
  • Molecular Weight: 150 kDa
  • Storage: Store at 4°C, do not freeze

Catalog No. 50-152-8430


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.