Learn More
Bio X Cell InVivoMAb anti-mouse CD20, MB20-11, IgG2c, 1 MG
Supplier: Bio X Cell BE0356R001MG
The MB20-11 monoclonal antibody targets mouse CD20, a 33-37 kDa transmembrane protein found on immature and mature B cells. It effectively depletes B cells via antibody-dependent cellular cytotoxicity (ADCC), making it valuable for B cell biology research and therapeutic development.
- Isotype: Mouse IgG2c, κ
- Immunogen: Mouse CD20-GFP transfected 300.19 cells
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: ≤1 EU/mg (≤0.001 EU/μg)
- Purity: ≥95%, determined by SDS-PAGE
- Sterility: 0.2 μm filtration
- Purification method: Protein A
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.