Learn More
Bio X Cell InVivoMAb, anti-mouse CD20, MB20-11, IgG2c, 100 MG
Supplier: Bio X Cell BE0356R100MG
The MB20-11 monoclonal antibody targets mouse CD20, a B cell-specific 33-37 kDa transmembrane protein crucial for B cell activation and immune response. It rapidly depletes B cells via antibody-dependent cellular cytotoxicity (ADCC) by engaging Fcγ receptors on monocytes.
- Reacts with mouse CD20
- Isotype: mouse IgG2c, κ
- Immunogen: mouse CD20-GFP transfected 300.19 cells
- Applications: in vivo B cell depletion, Western blot
- Formulation: PBS, pH 7.0, no stabilizers/preservatives
- Endotoxin: ≤1 EU/mg (≤0.001 EU/μg)
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Production: purified from animal-free cell culture supernatant
- Purification: protein A
- Molecular weight: 150 kDa
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.