Learn More
Bio X Cell anti-mouse CD20, Clone MB20-11, IgG2c, 25 MG
Supplier: Bio X Cell BE0356R025MG
The InVivoMAb anti-mouse CD20, clone MB20-11, is a monoclonal antibody that targets mouse CD20, a 33-37 kDa transmembrane protein expressed on immature and mature B cells. This antibody effectively depletes B cells via antibody-dependent cellular cytotoxicity (ADCC), making it a valuable tool for studying B cell biology and developing therapies.
- Reacts with mouse CD20
- Isotype: Mouse IgG2c, κ
- Applications: in vivo B cell depletion, Western blot
- Formulation: PBS, pH 7.0; contains no stabilizers or preservatives
- Purity: ≥95%, determined by SDS-PAGE
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Sterility: 0.2 μm filtration
- Production: Purified from cell culture supernatant in an animal-free facility
- Purification: Protein A
- Molecular weight: 150 kDa
- Storage: Store at stock concentration at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.