Learn More
Bio X Cell anti-mouse CD20, Clone MB20-11, 5 MG
Supplier: Bio X Cell BE0356R005MG
The MB20-11 monoclonal antibody targets mouse CD20, a B cell-specific transmembrane protein involved in intracellular calcium regulation and B cell activation. It effectively depletes B cells via antibody-dependent cellular cytotoxicity (ADCC), making it a valuable tool for studying B cell biology and developing therapies for autoimmune diseases and malignancies.
- Reacts with mouse CD20
- Depletes B cells through antibody-dependent cellular cytotoxicity (ADCC)
- Isotype: mouse IgG2c, κ
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Purity: ≥95% (determined by SDS-PAGE)
- Store at stock concentration at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.