Learn More
Bio X Cell Antibody, Mouse FasL, MFL3, 1 MG
Supplier: Bio X Cell BE0319R001MG
The MFL3 monoclonal antibody targets mouse Fas Ligand (FasL), also known as CD178 or CD95 Ligand, which is a 40 kDa type II transmembrane glycoprotein. This antibody is crucial for studying FasL-induced apoptotic cell death and its roles in peripheral tolerance, immune evasion by tumors, and neutrophil function.
- Reacts with mouse Fas Ligand (CD178)
- Isotype: Armenian hamster IgG
- Molecular weight: 150 kDa
- Immunogen: BHK cells expressing B6 mouse FasL
- Purification method: Protein A
- Purity: ≥95% by SDS-PAGE
- Endotoxin level: ≤1 EU/mg
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Sterility: 0.2 μm filtration
- Validated for in vivo/in vitro FasL blockade, functional assay, IHC (paraffin), and flow cytometry
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.