Learn More
Bio X Cell Monoclonal Antibody, MIAP301, IgG2a, Mouse, 1 MG
Supplier: Bio X Cell BE0270R001MG
The MIAP301 monoclonal antibody reacts with mouse CD47, also known as integrin-associated protein (IAP). CD47 is a 50 kDa glycosylated transmembrane protein, widely expressed on hematopoietic and non-hematopoietic cells, involved in apoptosis, proliferation, adhesion, and migration. The MIAP301 antibody neutralizes CD47 both in vivo and in vitro.
- Clone: MIAP301
- Host: Rat
- Target: Mouse CD47 (IAP)
- Reactivity: Mouse
- Applications: In vivo/in vitro CD47 blockade, Immunofluorescence
- Isotype: Rat IgG2a, κ
- Purification: Protein G from animal-free facility
- Formulation: PBS, pH 7.0, no stabilizers/preservatives, endotoxin ≤1EU/mg, purity ≥95%, 0.2 μm sterile filtered
- Storage: 4°C at stock concentration, do not freeze
- Molecular weight: 150 kDa
- Conjugation: Unconjugated
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.