Learn More
Bio X Cell Antibody, Human/Mouse/Rat Amyloid-beta, MOAB-2, IgG2b, 100 MG
Supplier: Bio X Cell BE0368R100MG
The MOAB-2 monoclonal antibody recognizes unaggregated, oligomeric, or fibrillar forms of Aβ42 and unaggregated Aβ40, showing selectivity for human Aβ42. It detects Aβ but not amyloid precursor protein (APP) in synthetic Aβ or human/rodent brain homogenates.
- Host species: mouse
- Isotype: mouse IgG2b, λ
- Immunogen: human Aβ42
- Purity: ≥95% (determined by SDS-PAGE)
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Sterility: 0.2 μm filtration
- Purification: protein A
- Formulation: PBS, pH 7.0 (contains no stabilizers or preservatives)
- Storage: store at stock concentration at 4°C; do not freeze
- Applications: western blot, immunohistochemistry, immunofluorescence, immunoprecipitation, ELISA
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.