Learn More
Bio X Cell Mouse IgG2a Fc, Purity 95, 1 MG
Supplier: Bio X Cell BE0097R001MG
This recombinant mouse IgG2a Fc is the Fc fragment of mouse IgG2a, commonly used as an isotype control for fusion proteins containing the mouse IgG Fc fragment. Its molecular mass is approximately 34 kDa in SDS-PAGE under reducing conditions.
- Purity ≥95% (determined by SDS-PAGE)
- Endotoxin level ≤1EU/mg (determined by LAL assay)
- Formulated in PBS, pH 7.0, without stabilizers or preservatives
- Purified from cell culture supernatant in an animal-free facility
- Sterility ensured by 0.2 μm filtration
- Purification method: Protein A
- Recommended dilution buffer: InVivoPure pH 7.0 Dilution Buffer
- Store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.