Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell Mouse IgG2a D265A Isotype Control, 0.25 MG

Catalog No. NC3365125
Encompass_Preferred

This mouse IgG2a isotype control antibody reacts with hen egg lysozyme and has low or no specific binding to any mouse sample. The D265A substitution has been shown to decrease effector functions by disrupting the Fc/Fcγ receptor interface.

  • Isotype: Mouse IgG2a
  • Recommended dilution buffer: pH 7.0 dilution buffer
  • Conjugation: Unconjugated
  • Mutations: D265A
  • Immunogen: Hen egg lysozyme (HEL)
  • Formulation: PBS, pH 7.0
  • Endotoxin: ≤0.5EU/mg (≤0.0005EU/μg)
  • Aggregation: <5%
  • Purity: ≥95%
  • Sterility: 0.2 μm filtration
  • Production: Purified from CHO cell supernatant
  • Purification: Protein A
  • RRID: AB_2927524
  • Molecular weight: 150 kDa
  • Storage: Store at 4°C; do not freeze

Catalog No. NC3365125 Supplier Bio X Cell Supplier No. CP150R025MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This mouse IgG2a isotype control antibody reacts with hen egg lysozyme and has low or no specific binding to any mouse sample. The D265A substitution has been shown to decrease effector functions by disrupting the Fc/Fcγ receptor interface.

  • Isotype: Mouse IgG2a
  • Recommended dilution buffer: pH 7.0 dilution buffer
  • Conjugation: Unconjugated
  • Mutations: D265A
  • Immunogen: Hen egg lysozyme (HEL)
  • Formulation: PBS, pH 7.0
  • Endotoxin: ≤0.5EU/mg (≤0.0005EU/μg)
  • Aggregation: <5%
  • Purity: ≥95%
  • Sterility: 0.2 μm filtration
  • Production: Purified from CHO cell supernatant
  • Purification: Protein A
  • RRID: AB_2927524
  • Molecular weight: 150 kDa
  • Storage: Store at 4°C; do not freeze

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.