Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell InVivoMAb Anti-Mouse CD154, Clone MR-1, 1 MG

Catalog No. 501528244
Encompass_Preferred

The MR-1 monoclonal antibody reacts with mouse CD154 (CD40 ligand), a 39 kDa accessory molecule belonging to the TNF superfamily of cytokines. It is primarily expressed on activated CD4+ T lymphocytes and signals through CD40, playing a crucial role in T and B lymphocyte costimulation.

  • Clone: MR-1
  • Isotype: Armenian hamster IgG
  • Immunogen: Activated mouse Th1 clone D1.6
  • Reported applications: in vivo blocking of CD40/CD40L signaling, in vitro blocking of CD40/CD40L signaling, Western blot, in vitro organoids/organ-on-chip
  • Formulation: PBS, pH 7.0; contains no stabilizers or preservatives
  • Endotoxin: ≤1EU/mg (≤0.001EU/µg)
  • Purity: ≥95%
  • Sterility: 0.2 µm filtration
  • Production: Purified from cell culture supernatant in an animal-free facility
  • Purification: Protein A
  • Storage: Store at 4°C; do not freeze

Catalog No. 50-152-8244 Supplier Bio X Cell Supplier No. BE00171R001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The MR-1 monoclonal antibody reacts with mouse CD154 (CD40 ligand), a 39 kDa accessory molecule belonging to the TNF superfamily of cytokines. It is primarily expressed on activated CD4+ T lymphocytes and signals through CD40, playing a crucial role in T and B lymphocyte costimulation.

  • Clone: MR-1
  • Isotype: Armenian hamster IgG
  • Immunogen: Activated mouse Th1 clone D1.6
  • Reported applications: in vivo blocking of CD40/CD40L signaling, in vitro blocking of CD40/CD40L signaling, Western blot, in vitro organoids/organ-on-chip
  • Formulation: PBS, pH 7.0; contains no stabilizers or preservatives
  • Endotoxin: ≤1EU/mg (≤0.001EU/µg)
  • Purity: ≥95%
  • Sterility: 0.2 µm filtration
  • Production: Purified from cell culture supernatant in an animal-free facility
  • Purification: Protein A
  • Storage: Store at 4°C; do not freeze

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.