Learn More
Bio X Cell Llama IgG, Polyclonal, Unconjugated, 1 MG
Supplier: Bio X Cell BE0369R001MG
The polyclonal llama IgG is purified from normal llama serum. Camelids produce antibodies that lack a light chain, known as heavy chain-only antibodies (HCAbs). Llama IgG purified by Protein G will primarily consist of IgG1 and IgG3.
- Purified from normal llama serum
- Contains heavy chain-only antibodies (HCAbs)
- Formulated in PBS, pH 7.0, without stabilizers or preservatives
- Purity ≥95% as determined by SDS-PAGE
- Sterility by 0.2 μm filtration
- Endotoxin level ≤1EU/mg (≤0.001EU/μg)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.