Learn More
Bio X Cell InVivoMAb Llama IgG, Polyclonal, Unconjugated, 100 MG
Supplier: Bio X Cell BE0369R100MG
This polyclonal llama IgG is purified from normal llama serum. Camelids produce heavy chain-only antibodies (HCAbs), which lack a light chain. Llama IgG2 and IgG3 are HCAbs, while IgG1 is a conventional IgG. Protein G purification primarily yields IgG1 and IgG3.
- Isotype: Llama IgG
- Conjugation: Unconjugated
- Formulation: PBS, pH 7.0; no stabilizers or preservatives
- Endotoxin: ≤1 EU/mg (≤0.001 EU/μg) by LAL assay
- Sterility: 0.2 μm filtration
- Purification: Protein G
- Purity: ≥95% by SDS-PAGE
- Molecular weight: 90-100 kDa (heavy chain-only antibodies); 150 kDa (complete IgG)
- Storage: Store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.