Learn More
Bio X Cell Llama IgG, Polyclonal, Purified, Unconjugated, 50 MG
Supplier: Bio X Cell BE0369R050MG
This polyclonal llama IgG is purified from normal llama serum. Camelids express antibodies lacking a light chain, known as heavy chain-only antibodies (HCAbs). Llama IgG2 and IgG3 are HCAbs, while IgG1 is conventional IgG with light and heavy chains.
- Isotype: Llama IgG
- Unconjugated
- Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/µg)
- Purity: ≥95%
- Sterility: 0.2 µm filtration
- Purification method: Protein G
- Molecular weight: 90-100 kDa (heavy chain-only antibodies), 150 kDa (complete IgG)
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.