Learn More
Bio X Cell InVivoMAb, mouse Ly6G/Ly6C, NIMP-R14, Unconjugated, 1 MG
Supplier: Bio X Cell BE0320R001MG
The NIMP-R14 monoclonal antibody is specific for murine neutrophils and reacts strongly with mouse Ly6G and Ly6C, previously known as GR-1. It is useful for depleting neutrophils in mice.
- Specific for murine neutrophils.
- Reacts strongly with mouse Ly6G and Ly6C.
- Isotype: Rat IgG2b, κ.
- Formulation: PBS, pH 7.0; contains no stabilizers or preservatives.
- Endotoxin: ≤1EU/mg (≤0.001EU/μg).
- Purity: ≥95%.
- Sterility: 0.2 μm filtered.
- Purified from cell culture supernatant in an animal-free facility by Protein A.
- Molecular weight: 150 kDa.
- Store at stock concentration at 4°C; do not freeze.
- Reported applications include in vivo neutrophil depletion, immunohistochemistry (paraffin and frozen), immunofluorescence, and flow cytometry.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.