Learn More
Bio X Cell InVivoMAb anti-mouse Ly6G/Ly6C, NIMP-R14, 100 MG
Supplier: Bio X Cell BE0320R100MG
The NIMP-R14 monoclonal antibody is specific for murine neutrophils, reacting strongly with mouse Ly6G and Ly6C. This antibody is useful for depleting neutrophils in mice and is involved in cell signaling and adhesion processes.
- Clone: NIMP-R14
- Reactivities: mouse
- Isotype: rat IgG2b, κ
- Immunogen: purified BALB/c mouse neutrophils
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95% by SDS-PAGE
- Sterility: 0.2 μm filtration
- Production: purified from cell culture supernatant in an animal-free facility
- Purification: protein A
- Molecular weight: 150 kDa
- Applications include in vivo neutrophil depletion, immunohistochemistry (paraffin & frozen), immunofluorescence, and flow cytometry.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.