Learn More
Bio X Cell InVivoMAb, anti-mouse CD38, Clone NIMR5, 1 MG
Supplier: Bio X Cell BE0317R001MG
This InVivoMAb anti-mouse CD38 antibody is a high-purity, low-endotoxin, preservative-free monoclonal antibody suitable for in vivo preclinical research. It is designed for applications such as in vivo and in vitro CD38 stimulation, in vitro B cell activation, and immunofluorescence.
- Clone: NIMR5
- Isotype: Rat IgG2a
- Suitable for in vivo CD38 stimulation
- Suitable for in vitro CD38 stimulation
- Suitable for in vitro B cell activation
- Suitable for immunofluorescence
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.