Learn More
Bio X Cell Anti-Mouse CD38, NIMR5, Rat IgG2a, 25 MG
Supplier: Bio X Cell BE0317R025MG
The NIMR5 monoclonal antibody reacts with mouse CD38, a 42 kDa type II transmembrane glycoprotein. CD38 is expressed on various immune cells and is involved in NAD homeostasis and intracellular calcium signaling. The NIMR5 antibody is agonistic, inducing B cell proliferation, protecting B cells from apoptosis, and slowing disease progression in tumor-bearing mice.
- Formulation: PBS, pH 7.0, with no stabilizers or preservatives
- Endotoxin: ≤1 EU/mg (≤0.001 EU/µg), determined by LAL assay
- Purity: ≥95%, determined by SDS-PAGE
- Sterility: 0.2 µm filtration
- Production: Purified from cell culture supernatant in an animal-free facility
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at stock concentration at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.