Learn More
Bio X Cell Anti-Mouse CD38 Antibody, Clone NIMR5, 50 MG
Supplier: Bio X Cell BE0317R050MG
The NIMR5 monoclonal antibody reacts with mouse CD38, a 42 kDa type II transmembrane glycoprotein expressed on various B and T cells. This agonistic antibody induces B cell proliferation, protects B cells from apoptosis, and slows disease progression in tumor-bearing mice.
- Agonistic antibody
- Reacts with mouse CD38
- Induces B cell proliferation and protects B cells from apoptosis
- Formulated in PBS, pH 7.0, without stabilizers or preservatives
- Endotoxin: ≤1EU/mg
- Purity: ≥95% by SDS-PAGE
- Sterility: 0.2 μm filtration
- Purified by Protein G
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.