Learn More
Bio X Cell Antibody, CD3 OKT-3, Mouse IgG2a, 1 MG
Supplier: Bio X Cell BE00012R001MG
This monoclonal antibody reacts with human CD3ε, a 20 kDa transmembrane cell-surface protein expressed on T lymphocytes, NK-T cells, and developing thymocytes. It plays a role in TCR signaling, T lymphocyte activation, and antigen recognition, with immunosuppressive properties in vivo.
- Clone OKT-3
- Reactivity with human CD3ε
- Isotype mouse IgG2a, κ
- Purified by protein G
- Formulated in PBS, pH 7.0, without stabilizers or preservatives
- Endotoxin level ≤1 EU/mg
- Purity ≥95% (SDS-PAGE)
- Sterile filtered to 0.2 μm
- Animal-free production
- Molecular weight 150 kDa
- Suitable for T cell stimulation/activation, T cell depletion, and flow cytometry
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.