Learn More
Bio X Cell Antibody, Human CD4 OKT-4, IgG2b, 1 MG
Supplier: Bio X Cell BE00032R001MG
InVivoMAb anti-human CD4 is a monoclonal antibody (Clone OKT-4) purified from cell culture supernatant. It is formulated in PBS, pH 7.0, without stabilizers or preservatives, making it suitable for various in vitro and in vivo applications.
- Reacts with human CD4
- Mouse IgG2b, κ isotype
- Purified by Protein G
- Endotoxin ≤1EU/mg
- Purity ≥95% by SDS-PAGE
- Sterility by 0.2 µm filtration
- Recommended for in vitro T cell stimulation/activation, in vivo CD4+ T cell depletion in humanized mice, flow cytometry, and immunoprecipitation
- Store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.