Learn More
Bio X Cell InVivoMAb Anti-Human CD71, Clone OKT9, 5 MG
Supplier: Bio X Cell BE0023R005MG
The OKT-9 monoclonal antibody reacts with human CD71 (Transferrin receptor protein 1, TfR1), a 95 kDa type II transmembrane glycoprotein. CD71 is expressed by immature proliferating cells and influences cellular proliferation by mediating iron uptake. It is considered a potential target in human leukemia and lymphoma.
- Target: human CD71 (transferrin receptor protein 1, TfR1)
- Clone: OKT-9
- Host species: mouse (isotype: mouse IgG1, κ)
- Reactivity: human
- Applications: immunohistochemistry (frozen), immunofluorescence, flow cytometry
- Purification: purified from cell culture supernatant using protein G
- Formulation: PBS, pH 7.0
- Endotoxin level: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95% by SDS-PAGE
- Molecular weight: 150 kDa
- Storage: store at 4°C. Do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.